Learn More
Glass Expansion Sampler ConeGuard PE NexION 1000/2000/5000/300/350 ICP-MS. Use to protect cone threads during cleaning.
SDP
Catalog No. 502703071
Encompass_Preferred
Sampler ConeGuard (N8145319). For use with PE NexION 1000/2000/5000/300/350 ICP-MS sampler cones to protect cone threads during cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3071 Supplier Glass Expansion Supplier No. 708031024
Sampler ConeGuard (N8145319). For use with PE NexION 1000/2000/5000/300/350 ICP-MS sampler cones to protect cone threads during cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001