Learn More
Hach Company Sampler, Dipper, Economy, 3ft
Catalog No. 501996825 Shop All Hach Company Products
Encompass
For convenient sampling from tanks, vats, streams, ponds, lakes, etc., these economy dippers are light, easy to use and portable. The polypropylene cups have threaded fittings that screw onto the handle. Two pouring spouts on the 500 mL (16 oz) dipper make it useful for left or right-handed people to pour from either side. Ends are sealed for easy cleaning and include a hook for hanging.
Catalog No. 50-199-6825 Supplier Hach Company Supplier No. 2929301
For convenient sampling from tanks, vats, streams, ponds, lakes, etc., these economy dippers are light, easy to use and portable. The polypropylene cups have threaded fittings that screw onto the handle. Two pouring spouts on the 500 mL (16 oz) dipper make it useful for left or right-handed people to pour from either side. Ends are sealed for easy cleaning and include a hook for hanging.