Learn More
Hach Company Extra Screw-on Cup for Sampler Dipper, 500 mL
Supplier:  Hach Company 2929401
Encompass
For convenient sampling from tanks, vats, streams, ponds, lakes, etc., these economy dippers are light, easy to use and portable. The polypropylene cups have threaded fittings that screw onto the handle. Two pouring spouts on the 500 mL (16 oz) dipper make it useful for left or right-handed people to pour from either side. Ends are sealed for easy cleaning and include a hook for hanging.
Catalog No. 50-199-6827