Learn More
Gentech Scientific Inc Liquid & Headspace Sampler for GC Systems
SDP
Catalog No. 501897509
Encompass_Preferred
Liquid- 121 position (2ml) liquid vial rack, Liquid Starter Kit: sample vials, caps, septa (10pcs), Liquid Kit: syringe adapter& spacers for solvent vials, Headspace- 42 Position (20ml) headspace vial rack, Headspace Starter Kit: 20ml vials, caps, septa (10pcs), 180 Day Warranty Different configurations are also available for liquid, headspace, and SPME. Compatible with all major manufacturer’s GC’s (Agilent, Thermo, Shimadzu and more) GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions.
Catalog No. 50-189-7509 Supplier Gentech Scientific Inc Supplier No. GTS11401
Add to Cart
Add to Cart
Request a Quote
Liquid- 121 position (2ml) liquid vial rack, Liquid Starter Kit: sample vials, caps, septa (10pcs), Liquid Kit: syringe adapter& spacers for solvent vials, Headspace- 42 Position (20ml) headspace vial rack, Headspace Starter Kit: 20ml vials, caps, septa (10pcs), 180 Day Warranty Different configurations are also available for liquid, headspace, and SPME. Compatible with all major manufacturer’s GC’s (Agilent, Thermo, Shimadzu and more) GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions.