Learn More
Trajan Scientific Sampler kit GAV 200 Gas 1/EA
SDP
Catalog No. 5000494423
Encompass_Preferred
Gas and vapor (GAV) sampling kit (contains canister bulb 10 sample bags marker pen and instructions)
Catalog No. 50-004-94423 Supplier Trajan Scientific Supplier No. 090111
Gas and vapor (GAV) sampling kit (contains canister bulb 10 sample bags marker pen and instructions)