Learn More
BURKLE INC Sampler Liquid CupSampler, V4A, 3.38 oz. (100 ml), 39.37 in. (100 cm)
Catalog No. 5000025792
Encompass_Preferred
It is possible to carry out point sampling of liquids of varying viscosity with the stainless steel AISI 316L (1.4404) Liquid CupSampler. Media-containing particles can also be sampled without difficulty.
The sampler can be used in the fields of chemistry, foodstuffs and cosmetics.
The cup can be unscrewed from the stem, allowing easy cleaning.
Catalog No. 50-000-25792 Supplier BURKLE INC Supplier No. 53570001
It is possible to carry out point sampling of liquids of varying viscosity with the stainless steel AISI 316L (1.4404) Liquid CupSampler. Media-containing particles can also be sampled without difficulty.
The sampler can be used in the fields of chemistry, foodstuffs and cosmetics.
The cup can be unscrewed from the stem, allowing easy cleaning.