Learn More
BURKLE INC Sampler Tubus, V4A, 15.75 in. (40 cm), Ø 1.57 in. (40 mm), ISTA standard
Catalog No. 5000025642
Encompass_Preferred
Sampler for pourable bulk goods up to approx. 1 cm in diameter. This makes it possible, for example, to remove hazel nuts, blends of tea, grain or similar items from large sacks.
Catalog No. 50-000-25642 Supplier BURKLE INC Supplier No. 53820040
Sampler for pourable bulk goods up to approx. 1 cm in diameter. This makes it possible, for example, to remove hazel nuts, blends of tea, grain or similar items from large sacks.