Learn More
Hach Company Sampler, Dipper
Supplier:  Hach Company 2929501
Encompass
For taking samples of water, sewage, wastes and effluents from lakes, streams, tanks, or ponds, the water sample dipper provides a long reach for otherwise inaccessible spots. A 1000 mL graduated polypropylene pitcher is mounted on an aluminum tube handle. The handle has 3 parts, a 91cm (36") lower section, a 112cm (44") telescoping section, and an additional 91cm (36") upper section. Connections are made with spring buttons with adjusting steps, 15cm (72") apart. Minimum handle length is 91cm (36"), maximum length is 274cm (108"). Handle diameter is 25mm (1") with 22mm (7/8") telescoping tube.
Catalog No. 50-199-6828