Learn More
BURKLE INC Sampling drill for stalk material, Ø 1.77 in (45 mm), 15.75 in (400 mm)
Supplier:  BURKLE INC 53374040
Encompass_Preferred
The stalk sampler is a sampler designed specifically for hay, silage, straw, herbs, shrubs or other dry, stalky plant materials where other samplers quickly become tangled. The special geometry of the auger tip cuts effectively and quickly into various stalky materials.
The hay and silage sampling auger takes samples from a silo or bale quickly, cleanly and easily. Sampling with mechanical assistance from a drill or powerful cordless screwdriver allows for effortless removal of the sample with little manual effort.
The auger can be used vertically (sampling from the top) or horizontally (sampling from the side).
The rugged and durable tube auger is made entirely of stainless steel 316L (1.4404)/1.4034 or aluminum/stainless steel 1.4034.
The handy ejector assists in emptying the sample from the auger.
Catalog No. 50-000-25653