Learn More
BURKLE INC Ladle, long handle, 1.01 oz. (30 ml), Bio-PE
Catalog No. 503081211
Encompass_Preferred
The sampler made of Green PE is produced in our class 7 (10,000) clean room and individually packaged for disposable use.
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates. The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container.
A long handle with indentations for fingers ensures a firm hold and precise work.
Catalog No. 50-308-1211 Supplier BURKLE INC Supplier No. 53790026
The sampler made of Green PE is produced in our class 7 (10,000) clean room and individually packaged for disposable use.
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates. The ladle has a very smooth bottom and rounded corners. This makes it possible to reach into any corner of a container or to take medium directly from the bottom of the container.
A long handle with indentations for fingers ensures a firm hold and precise work.