Learn More
BURKLE INC Palette knife spatula, LxW 7.56 x 0.79 in (192 x 20 mm), Bio-PE
Catalog No. 503081212
Encompass_Preferred
The sampler made of Green PE is produced in our class 7 (10,000) clean room and individually packaged.
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The sampling palette knife spatula can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The palette knife spatula reaches easily into the corners and curves of various containers and is suitable for scraping out bowls, removing residue or wiping off Bürkle's volumetric and dosing spoons.
Catalog No. 50-308-1212 Supplier BURKLE INC Supplier No. 53790035
The sampler made of Green PE is produced in our class 7 (10,000) clean room and individually packaged.
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The sampling palette knife spatula can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The palette knife spatula reaches easily into the corners and curves of various containers and is suitable for scraping out bowls, removing residue or wiping off Bürkle's volumetric and dosing spoons.