Learn More
BURKLE INC Sampling scoop, long handle, 1.69 oz. (50 ml), Bio-PE
Catalog No. 503081273
Encompass_Preferred
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids.
The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.
Catalog No. 50-308-1273 Supplier BURKLE INC Supplier No. 53790024
Green PE bio-plastic has all the suitable properties of conventional polyethylene but is manufactured from renewable raw materials and can be completely recycled.
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids.
The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.