Learn More
Research Products International Corp Sampling Scp S Lrg 250ml 10pk
SDP
Catalog No. 502388434
Encompass
Durable Sampling Scoops are designed for easy sample retrieval of granules, powders and samples.Disposable,sterile scoops are idea for single use and prevent cross-contamination. Scoops feature large, wide openings for easy sample retrieval; flat bottom to stabilize sample on scale;shatterproof, BSE/TSE free. Maufactured from FDA food grade polystyrene in an ISO8 Class clean room; meet quality control and GMP requirements for sampling.
Durable design
Sterile, Individually wrapped
Safe sample recovery
Catalog No. 50-238-8434 Supplier Research Products International Corp Supplier No. 249511
Durable Sampling Scoops are designed for easy sample retrieval of granules, powders and samples.Disposable,sterile scoops are idea for single use and prevent cross-contamination. Scoops feature large, wide openings for easy sample retrieval; flat bottom to stabilize sample on scale;shatterproof, BSE/TSE free. Maufactured from FDA food grade polystyrene in an ISO8 Class clean room; meet quality control and GMP requirements for sampling.
Durable design
Sterile, Individually wrapped
Safe sample recovery