Learn More
Sigma Aldrich Fine Chemicals Biosciences Sampling spatula polypropy
Catalog No. 502900885
Encompass
One-piece spatula is molded from polypropylene and retains approximately 15cc of material. Shape allows easy transfer of sample to container. Use to sample directly from production line or containers including bags and drums.V-shaped spatula is perfect for sampling granulated or semi solid material Non-sterile Use different spatula for different materials to reduce cross contamination Sharp point will pierce most packaging
Catalog No. 50-290-0885 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BAF37840000012EA
One-piece spatula is molded from polypropylene and retains approximately 15cc of material. Shape allows easy transfer of sample to container. Use to sample directly from production line or containers including bags and drums.V-shaped spatula is perfect for sampling granulated or semi solid material Non-sterile Use different spatula for different materials to reduce cross contamination Sharp point will pierce most packaging