Learn More
BURKLE INC Sampling spoon, long handle, 0.17 oz. (5 ml), PS, white
Catalog No. 503081255
Encompass_Preferred
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids.
The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.
Catalog No. 50-308-1255 Supplier BURKLE INC Supplier No. 53785001
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids.
The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.