Learn More
Chemglass Life Sciences Sampling Tube, Reduced Diameter, M6 Thread, 1/8"OD Tubing, for 2-5L Bioreactors, 210mm Max Immersion Length
SDP
Catalog No. 501294431
Encompass
Sampling tube with a reduced diameter which requires a smaller amount of sample to be drawn from the reactor.Tubing is 1/8 inch OD.Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc.Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
This item is not returnable. View return policy
Catalog No. 50-129-4431 Supplier Chemglass Life Sciences Supplier No. CLS1380R33
Sampling tube with a reduced diameter which requires a smaller amount of sample to be drawn from the reactor.Tubing is 1/8 inch OD.Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc.Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.