Learn More
Cayman Chemical Sampling tubes with BHT and I
SDP
Catalog No. 502968057
Encompass_Preferred
These tubes are specifically designed to be used for 1 ml blood samples which will be assayed both for 8-isoprostanes and prostaglandins. BHT sampling tubes may be useful for any lipid peroxidation and COX metabolites studies.
Catalog No. 50-296-8057 Supplier Cayman Chemical Supplier No. 1853435 EA
These tubes are specifically designed to be used for 1 ml blood samples which will be assayed both for 8-isoprostanes and prostaglandins. BHT sampling tubes may be useful for any lipid peroxidation and COX metabolites studies.