Learn More
Cayman Chemical Samplng Tubes wIth PHMB 35ea
SDP
Catalog No. 502951527
Encompass_Preferred
4-(hydroxymercuri)Benzoic Acid(PHMB) is used to inhibit cysteine-protease and acts as a reversible thiol-specific blocker. While blocking the acylated ghrelin degradation, it allows the AcetylCholine Esterase AChE™ used in SPI-Bio EIA kits to work normally, in contrast to serine-protease inhibitors.
Catalog No. 50-295-1527 Supplier Cayman Chemical Supplier No. 1094935 EA
4-(hydroxymercuri)Benzoic Acid(PHMB) is used to inhibit cysteine-protease and acts as a reversible thiol-specific blocker. While blocking the acylated ghrelin degradation, it allows the AcetylCholine Esterase AChE™ used in SPI-Bio EIA kits to work normally, in contrast to serine-protease inhibitors.