Learn More
Cayman Chemical Samplng tubS W BHT nd I 150ea
SDP
Catalog No. 502965262
Encompass_Preferred
These tubes are specifically designed to be used for 1 ml blood samples which will be assayed both for 8-isoprostanes and prostaglandins. BHT sampling tubes may be useful for any lipid peroxidation and COX metabolites studies.
Catalog No. 50-296-5262 Supplier Cayman Chemical Supplier No. 18534150 EA
These tubes are specifically designed to be used for 1 ml blood samples which will be assayed both for 8-isoprostanes and prostaglandins. BHT sampling tubes may be useful for any lipid peroxidation and COX metabolites studies.