Learn More
Cayman Chemical Samplng TubS W Indomtha 100ea
SDP
Catalog No. 502951198
Encompass_Preferred
Indomethacin is a COX inhibitor that prevents ex vivo TXB2 formation and other PGs that can not be prevented using anticoagulant alone. The samples have to be transferred in indomethacin sampling tubes as soon as possible after blood collection.
Catalog No. 50-295-1198 Supplier Cayman Chemical Supplier No. 10951100 EA
Indomethacin is a COX inhibitor that prevents ex vivo TXB2 formation and other PGs that can not be prevented using anticoagulant alone. The samples have to be transferred in indomethacin sampling tubes as soon as possible after blood collection.