Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITL BioMedical SampLok™ Sampling Kit

Catalog No. 22799908 Shop All ITL BioMedical Products
Change view
Click to view available options
Volume (Metric):
10 mL
16 mL
20 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-799-908 10 mL
22-799-907 16 mL
22-799-909 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-799-908 Supplier ITL BioMedical Supplier No. A100750
Add to Cart
Add to Cart
This item is not returnable. View return policy

SampLok™ Sampling Kit provides a more straightforward, safer, and efficient way to collect and transfer platelet samples for bacterial testing.

  • Reduces the number of procedure steps, streamlining and simplifying the collection and transfer of biological samples
  • SampLok™ Sampling Kit can be used with various culture bottles and vacuum tubes
  • Protect staff from needlestick injuries

Specifications

Product Type Collection and Transport System
Format Tube
Sterility Sterile
Closure Type Inegrated Safety Lid
For Use With (Equipment) Culture Bottles and Vacuum Tubes
Quantity 110 per Case
Volume (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.