Learn More
Enzo Life Sciences SAMS peptide (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP2310001
Encompass_Preferred
This peptide, HMRSAMSGLHLVKRR, is derived from rat acetyl-CoA carboxylase and is known to be a good substrate for AMPK. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1299