Learn More
Enzo Life Sciences SAMS peptide (biotinylated) (1 mg)
SDP
Supplier:  Enzo Life Sciences BMLP2320001
Encompass_Preferred
AMPK substrate. This peptide, biotin-HMRSAMSGLHLVKRR, contains a biotin group at its amino terminus. It is derived from rat acetyl-CoA carboxylase and is known to be a good substrate for AMPK. The biotin allows peptide to be used in kinase assays with streptavidin-bound membranes. MW: 2118. Purity: ≥95%. Sequence biotin-His-Met-Arg-Ser-Ala-Met-Ser-Gly-Leu-His-Leu-Val-Lys-Arg-Arg-OH. Long Term Storage: -20°C
Catalog No. NC1475096