Learn More
Aviva Systems Biology Corp SAMSN1 Antibody - middle region (ARP45874_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45874_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAMSN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SAMSN1. Alternate names: HACS1, NASH1, SLy2, SASH2, SH3D6BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-222