Learn More
Medchemexpress LLC HY-103712A 10mg , Samuraciclib (hydrochloride) CAS:1805789-54-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103712A10MG
Encompass_Preferred
HY-103712A 10mg Samuraciclib (hydrochloride) CAS: 1805789-54-1 Samuraciclib hydrochloride (CT7001 hydrochloride) is a potent, selective, ATP-competitive and orally active CDK7 inhibitor, with an IC50 of 41 nM. Samuraciclib hydrochloride displays 45-, 15-, 230- and 30-fold selectivity over CDK1, CDK2 (IC50 of 578 nM), CDK5 and CDK9, respectively. Samuraciclib hydrochloride inhibits the growth of breast cancer cell lines with GI50 values between 0.2-0.3 µM. Samuraciclib hydrochloride has anti-tumor effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0453