Learn More
Medchemexpress LLC Samuraciclib hydroch 25mg | 1805789-54-1 | 25MG
SDP
Catalog No. 5000243233
Encompass_Preferred
Samuraciclib hydrochloride (CT7001 hydrochloride) is a potent selective ATP-competitive and orally active CDK7 inhibitor with an IC50 of 41 nM Samuraciclib hydrochloride displays 45- 15- 230- and 30-fold selectivity over CDK1 CDK2 (IC50 of 578 nM) CDK5 and CDK9 respectively Samuraciclib hydrochloride inhibits the growth of breast cancer cell lines with GI50 values between 0 2-0 3 M Samuraciclib hydrochloride has anti-tumor effects[1 [2
Catalog No. 50-002-43233 Supplier Medchemexpress LLC Supplier No. HY103712A25MG
Samuraciclib hydrochloride (CT7001 hydrochloride) is a potent selective ATP-competitive and orally active CDK7 inhibitor with an IC50 of 41 nM Samuraciclib hydrochloride displays 45- 15- 230- and 30-fold selectivity over CDK1 CDK2 (IC50 of 578 nM) CDK5 and CDK9 respectively Samuraciclib hydrochloride inhibits the growth of breast cancer cell lines with GI50 values between 0 2-0 3 M Samuraciclib hydrochloride has anti-tumor effects[1 [2