Learn More
TARGETMOL CHEMICALS INC SAMURACICLIB HYDROCHLORI 5MG
SDP
Catalog No. 502581622
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Samuraciclib hydrochloride (ICEC0942 hydrochloride) is a potent selective ATP competitive and oral active CDK7 inhibitor with IC50 of 41 nM. The selectivity of Samuraciclib hydrochloride is 45- 15- 230- and 30-fold higher than CDK1 CDK2 (IC50 is 578 nM) CDK5 and CDK9 respectively. Samuraciclib hydrochloride inhibits the growth of breast cancer cell lines with GI50 values between 0.2-0.3 uM. Samuraciclib hydrochloride has anti-tumor effects. purity: 99%
Catalog No. 50-258-1622 Supplier TARGETMOL CHEMICALS INC Supplier No. T108985MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Samuraciclib hydrochloride (ICEC0942 hydrochloride) is a potent selective ATP competitive and oral active CDK7 inhibitor with IC50 of 41 nM. The selectivity of Samuraciclib hydrochloride is 45- 15- 230- and 30-fold higher than CDK1 CDK2 (IC50 is 578 nM) CDK5 and CDK9 respectively. Samuraciclib hydrochloride inhibits the growth of breast cancer cell lines with GI50 values between 0.2-0.3 uM. Samuraciclib hydrochloride has anti-tumor effects. purity: 99%