Learn More
Medchemexpress LLC Samuraciclib hydrochloride | 1805789-54-1 | MFCD31657343 | 99.9% | 430.97 g/mol | C22H31ClN6O | 100 MG
SDP
Catalog No. 5000243860
Encompass_Preferred
Samuraciclib hydrochloride is a potent, selective, ATP-competitive, and orally active inhibitor of cyclin-dependent kinase 7 (CDK7) used in preclinical research. It inhibits proliferation of cancer cell lines and has demonstrated anti-tumor activity in vitro, making it a tool compound for studying CDK7-related transcriptional control and apoptosis pathways.
  • Potent, selective CDK7 inhibition (IC50 ~41 nM).
  • Orally active compound suitable for in vitro and preclinical studies.
  • Shown to inhibit cancer cell growth (GI50 0.2-0.3 μM in breast cancer lines).
  • Applicable for research on transcriptional regulation and apoptosis mechanisms.
  • Supplied as a stable solid with recommended refrigerated storage to maintain integrity.
Catalog No. 50-002-43860 Supplier Medchemexpress LLC Supplier No. HY103712A100MG
Samuraciclib hydrochloride is a potent, selective, ATP-competitive, and orally active inhibitor of cyclin-dependent kinase 7 (CDK7) used in preclinical research. It inhibits proliferation of cancer cell lines and has demonstrated anti-tumor activity in vitro, making it a tool compound for studying CDK7-related transcriptional control and apoptosis pathways.
  • Potent, selective CDK7 inhibition (IC50 ~41 nM).
  • Orally active compound suitable for in vitro and preclinical studies.
  • Shown to inhibit cancer cell growth (GI50 0.2-0.3 μM in breast cancer lines).
  • Applicable for research on transcriptional regulation and apoptosis mechanisms.
  • Supplied as a stable solid with recommended refrigerated storage to maintain integrity.