Learn More
Medchemexpress LLC Samuraciclib hydrochloride hydrate | 00-00-0 | 99.9% | 552.33 | C22H33ClN6O2 | 5 MG
SDP
Catalog No. 5000243736
Encompass_Preferred
Samuraciclib hydrochloride hydrate is the hydrochloride hydrate salt of samuraciclib, a potent, selective, ATP-competitive, orally active CDK7 inhibitor used for research. It inhibits CDK7 with an IC50 of 41 nM, shows selectivity versus related cyclin-dependent kinases, and reduces proliferation of breast cancer cell lines (GI50 0.2-0.3 μM).
  • Potent CDK7 inhibition (IC50 41 nM).
  • Selective versus CDK1, CDK2, CDK5, and CDK9.
  • Demonstrated cellular activity in breast cancer cell lines (GI50 0.2-0.3 μM).
  • High chemical purity (99.88%).
  • Hydrochloride hydrate salt form for improved handling.
  • Available in small research pack sizes suitable for in vitro studies.
Catalog No. 50-002-43736 Supplier Medchemexpress LLC Supplier No. HY103712C5MG
Samuraciclib hydrochloride hydrate is the hydrochloride hydrate salt of samuraciclib, a potent, selective, ATP-competitive, orally active CDK7 inhibitor used for research. It inhibits CDK7 with an IC50 of 41 nM, shows selectivity versus related cyclin-dependent kinases, and reduces proliferation of breast cancer cell lines (GI50 0.2-0.3 μM).
  • Potent CDK7 inhibition (IC50 41 nM).
  • Selective versus CDK1, CDK2, CDK5, and CDK9.
  • Demonstrated cellular activity in breast cancer cell lines (GI50 0.2-0.3 μM).
  • High chemical purity (99.88%).
  • Hydrochloride hydrate salt form for improved handling.
  • Available in small research pack sizes suitable for in vitro studies.