Learn More
MSC SAMURAI C/R GLVS
Catalog No. 5000788625
Encompass
The CR18NFT is a high visibility, ultra-light cut resistant glove designed for industrial protection. It features an 18 gauge Tuffalene HDPE shell and a New Foam Technology Nitrile dipped palm, offering comfort, dexterity, and breathability for a variety of applications.
  • High visibility ultra-light 18 gauge design
  • Tuffalene HDPE liner
  • New Foam Technology Nitrile dipped palm
  • Excellent dry, wet, and oily grip
  • Excellent tactile dexterity and cut protection
  • Machine washable
  • ANSI level A2 cut resistance
  • EN 388 levels: 4342
Catalog No. 50-007-88625 Supplier MSC Supplier No. 33679796
The CR18NFT is a high visibility, ultra-light cut resistant glove designed for industrial protection. It features an 18 gauge Tuffalene HDPE shell and a New Foam Technology Nitrile dipped palm, offering comfort, dexterity, and breathability for a variety of applications.
  • High visibility ultra-light 18 gauge design
  • Tuffalene HDPE liner
  • New Foam Technology Nitrile dipped palm
  • Excellent dry, wet, and oily grip
  • Excellent tactile dexterity and cut protection
  • Machine washable
  • ANSI level A2 cut resistance
  • EN 388 levels: 4342