Learn More
MSC Samurai Glove Gloves, ANSI A2 Cut Resistant, Tuffalene Platinum, 12/Pk
Catalog No. 5000673047
Encompass
This lightweight seamless glove features a salt and pepper 18-gauge Tuffalene® Platinum liner. The innovative Tuffalene® Platinum provides extra comfort, protection, and durability without the need for glass fiber or steel enhancements to achieve its level of cut protection. The shell is further enhanced by a black Mach Finish nitrile dipped palm.
  • ANSI Cut Level A2
  • EN 388 levels: 4X42B
  • Features an 18-gauge Tuffalene® Platinum UHMWPE shell, offering bare hand sensitivity and cut resistance, free from glass fiber or steel enhancements
  • Mach Finish nitrile palm is excellent for grip in wet and oily situations, and provides abrasion resistance
  • Knit wrist cuff prevents gloves from slipping off during work
Catalog No. 50-006-73047 Supplier MSC Supplier No. 30112114
This lightweight seamless glove features a salt and pepper 18-gauge Tuffalene® Platinum liner. The innovative Tuffalene® Platinum provides extra comfort, protection, and durability without the need for glass fiber or steel enhancements to achieve its level of cut protection. The shell is further enhanced by a black Mach Finish nitrile dipped palm.
  • ANSI Cut Level A2
  • EN 388 levels: 4X42B
  • Features an 18-gauge Tuffalene® Platinum UHMWPE shell, offering bare hand sensitivity and cut resistance, free from glass fiber or steel enhancements
  • Mach Finish nitrile palm is excellent for grip in wet and oily situations, and provides abrasion resistance
  • Knit wrist cuff prevents gloves from slipping off during work