Learn More
AdipoGen Sancycline
SDP
Supplier:  AdipoGen CDXS0344M250
Encompass_Preferred
Chemical. CAS 808-26-4. Formula C21H22N2O7. MW 414.4. Sancycline is a semisynthetic tetracycline antibiotic that is more active than tetracycline against 339 strains of anaerobic bacteria average MIC90s = 1 and 32 µg/ml, respectively. Sancycline is active against tetracycline-resistant E. coli, S. aureus, and E. faecalis strains with MICs ranging from 0.06 to 1 µg/ml. In vivo, sancycline is active against S. aureus in mice with ED50 values of 0.46 and 0.6 mg/kg for intravenous and subcutaneous administration, respectively. Like other tetracyclines, sancycline acts by reversibly binding to the 30S ribosomal subunit and inhibiting protein translation by blocking entry of aminoacyl-tRNA into the ribosome A site.
Catalog No. 50-256-0056