Learn More
TARGETMOL CHEMICALS INC SANCYCLINE 25MG
SDP
Catalog No. 502577014
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sancycline (Bonomycin) is a rare semi-synthetic tetracycline acts by reversibly binding to the 30S ribosomal subunit and inhibiting protein translation by blocking entry of aminoacyl-tRNA into the ribosome A site. purity: 99%
Catalog No. 50-257-7014 Supplier TARGETMOL CHEMICALS INC Supplier No. T305225MG
Add to Cart
Add to Cart
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sancycline (Bonomycin) is a rare semi-synthetic tetracycline acts by reversibly binding to the 30S ribosomal subunit and inhibiting protein translation by blocking entry of aminoacyl-tRNA into the ribosome A site. purity: 99%