Learn More
Andwin Scientific SAND FOR SAND BATH 1LB
SDP
Catalog No. NC9940882
Encompass_Preferred
The sand is a refill for the Modular Heating Sand Bath, a product that is ideal for heating blocks and irregular vessels. The sand is sold in a quantity of 454 grams or 1 pound.
Catalog No. NC9940882 Supplier Andwin Scientific Supplier No. 13259272
The sand is a refill for the Modular Heating Sand Bath, a product that is ideal for heating blocks and irregular vessels. The sand is sold in a quantity of 454 grams or 1 pound.