Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sand, Washed and Dried, Macron Fine Chemicals™

Catalog No. MK7062212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-7062-212 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7062-212 Supplier Avantor Supplier No. 706206

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14808-60-7
Physical Form Fine Granules
Molecular Formula O2Si
Formula Weight 60.08
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
Chemical Name or Material Sand

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Color Gray to White
Quantity 2.5 kg
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
Flash Point 0°C (32°F)
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
PubChem CID 24261
ChEBI CHEBI:30563
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.