Description
- Weights fill the Weighted Ballast Bag to 45 lb. to help maintain system stability
- Made of sand
- Provides additional ground support for the Barrier System
- Options and Accessories
- FS882: Weighted Ballast Bag
- FS871: 1000 Barrier System
- FS873: Quad Barrier System
- FS874: Octo Barrier System
- FS785: Copper Barrier System
Specifications
Specifications
| Product Type | Sand Weights |
| Material | Sand |