Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sand, White Quartz, Solstice

Catalog No. 60047651 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-651 5 kg Poly bottle
60-047-650 1 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-651 Supplier Solstice Supplier No. 833405KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purum p.a., powder

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2210°C
Melting Point 1710°C
Color White
Physical Form Solid
Quantity 5 kg
Odor Odorless
Packaging Poly bottle
Linear Formula SiO2
UN Number NONH for all modes of transport
Formula Weight 60.08g/mol
Chemical Name or Material Sand, white quartz
Show More Show Less
Hazard Statement P314
RTECSNumber 238-878-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.