Learn More
Grainger Sanding Disc 2 In 60G PK100
Catalog No. 502768106
Encompass_Preferred
The CARBORUNDUM Quick Change Disc, model 369TE4, is a 4-1/2 inch diameter x 7/8 inch thick Type 27 depressed center grinding wheel made of aluminum oxide abrasive grains bonded with vitrified bond. It has a maximum safe speed of 6600 RPM and is designed for use on portable grinders, stationary tools, and flexible shaft machines for general purpose grinding of ferrous metals. This thin wheel allows grinding in tight areas. It features a Quick Change hub system for fast and easy disc changes.
Catalog No. 50-276-8106 Supplier Grainger Supplier No. 16Y818
The CARBORUNDUM Quick Change Disc, model 369TE4, is a 4-1/2 inch diameter x 7/8 inch thick Type 27 depressed center grinding wheel made of aluminum oxide abrasive grains bonded with vitrified bond. It has a maximum safe speed of 6600 RPM and is designed for use on portable grinders, stationary tools, and flexible shaft machines for general purpose grinding of ferrous metals. This thin wheel allows grinding in tight areas. It features a Quick Change hub system for fast and easy disc changes.