Learn More
Electron Microscopy Sciences Flexible Shaft Machine Sanding Discs, Fine Grit, 3/4" diameter
SDP
Catalog No. 50996836
Encompass_Preferred
Yellow abrasive with press on/flip-off center hole feature. Aluminum oxide grit mounted on a thin backing, provides outstanding results in sanding and smoothing operations. Sold 100 discs in a tube.
Grit: Fine
Diameter: 3/4"
Catalog No. 50-996-836 Supplier Electron Microscopy Sciences Supplier No. 6214530
Yellow abrasive with press on/flip-off center hole feature. Aluminum oxide grit mounted on a thin backing, provides outstanding results in sanding and smoothing operations. Sold 100 discs in a tube.
Grit: Fine
Diameter: 3/4"