Learn More
Electron Microscopy Sciences Flexible Shaft Machine Sanding Discs, Medium Grit, 7/8" diameter
SDP
Catalog No. 50996835
Encompass_Preferred
Yellow abrasive with press on/flip-off center hole feature. Aluminum oxide grit mounted on a thin backing, provides outstanding results in sanding and smoothing operations. Sold 100 discs in a tube.
Grit: Medium
Diameter: 7/8"
Catalog No. 50-996-835 Supplier Electron Microscopy Sciences Supplier No. 6214521
Yellow abrasive with press on/flip-off center hole feature. Aluminum oxide grit mounted on a thin backing, provides outstanding results in sanding and smoothing operations. Sold 100 discs in a tube.
Grit: Medium
Diameter: 7/8"