Learn More
AVANTOR MASTERFLEX (VWR LLC) Warren Rupp S07B1K1KPNS000. Air-Powered Double-Diaphragm PVDF Pump, Santoprene; 23 GPM
Catalog No. NC1467033
Encompass
These pumps can move high-viscosity fluids such as paint, sludge, and sewage. However, flow rate and pressure depends on the application. If you have questions about the compatibility of your high-viscosity fluid, call our Application Specialists for more information. Order air filters/regulators to remove impurities from your air line and to help regulate working pressure. Combination air filter/regulators feature polypropylene filter, 0- to 160-psi gauge, and a polycarbonate bowl. Temperature range is 10 to 210¿F (-12 to 98¿C).
Catalog No. NC1467033 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7400021
These pumps can move high-viscosity fluids such as paint, sludge, and sewage. However, flow rate and pressure depends on the application. If you have questions about the compatibility of your high-viscosity fluid, call our Application Specialists for more information. Order air filters/regulators to remove impurities from your air line and to help regulate working pressure. Combination air filter/regulators feature polypropylene filter, 0- to 160-psi gauge, and a polycarbonate bowl. Temperature range is 10 to 210¿F (-12 to 98¿C).