Learn More
Kuhner Shaker Inc SANDWICH COVER FOR 24 DEEPWELL
Supplier:  Kuhner Shaker Inc SMCR1224G
Encompass
Sandwich cover for 24 deepwell MTPs CR1424a, b,c, and d..(with solid silicone, fixed pins, 1.2mm holes, low evap)
Catalog No. 50-257-0160