Learn More
Kuhner Shaker Inc SANDWICH COVER FOR 24 DEEPWELL
Catalog No. NC1057496
Encompass
Sandwich cover for all types of 24 deepwell MTPs, (low-evaporation version of CR1224, 1.2 mm holes). 104115.
Catalog No. NC1057496 Supplier Kuhner Shaker Inc Supplier No. SMCR1224A
Sandwich cover for all types of 24 deepwell MTPs, (low-evaporation version of CR1224, 1.2 mm holes). 104115.