Learn More
Kuhner Shaker Inc SANDWICH COVER FOR 96 DEEPWELL
Catalog No. NC1528567
Encompass
Sandwich cover for all types of 96 deepwell MTPs, (with solid silicone, 0.8 mm holes, low evaporation). 104118.
Catalog No. NC1528567 Supplier Kuhner Shaker Inc Supplier No. SMCR1296A
Sandwich cover for all types of 96 deepwell MTPs, (with solid silicone, 0.8 mm holes, low evaporation). 104118.