Learn More
Kuhner Shaker Inc SANDWICH COVER FOR ALL TYPES
Supplier:  Kuhner Shaker Inc SMCR1224E
Encompass
Sandwich cover for 24 deepwell MTP's, 18mm spacing, solid black silicone, 1.2mm holes low evap
Catalog No. NC1841548