Learn More
Kuhner Shaker Inc SANDWICH COVER FOR EXTRA DEEP
Supplier:  Kuhner Shaker Inc SMCR1224H
Encompass
Sandwich cover for extra deep 24 well plates (25ml each). Spongy silicone, 2.5mm holes
Catalog No. 50-257-0159