Learn More
Kuhner Shaker Inc SANDWICH COVER FOR EXTRA DEEP
Catalog No. 502570158
Encompass
Sandwich cover for extra deep 24 well plates (25ml per well). Spongey silicone, 1.2mm (small) holes
Catalog No. 50-257-0158 Supplier Kuhner Shaker Inc Supplier No. SMCR1224I
Sandwich cover for extra deep 24 well plates (25ml per well). Spongey silicone, 1.2mm (small) holes