Learn More
Kuhner Shaker Inc SANDWICH COVER FOR GREINER 96
Supplier:  Kuhner Shaker Inc SMCR1296C
Encompass
Sandwich cover for Greiner 96 deepwell MTPs and SMCR1496a, solid silicone, fixed with pins, 0.8 mm holes, low evap.
Catalog No. 50-257-0156