Learn More
Aqua Phoenix™ Sandy Situation Deluxe Kit, Safe Racer 2.0 Deluxe Kit Components
Shop All Fisherbrand Products

Description
Specifications
Specifications
| Grade | K to 5 |
| For Use With (Application) | Education Kit |
| Material | Plastic |
| Size | Medium weight |
| Product Type | Plastic fork |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.