Learn More
Medchemexpress LLC Sanfetrinem (sodium) | 141611-76-9 | 303.29 g/mol | C14H18NNaO5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY106922A10MM 1ML
Encompass_Preferred
Sanfetrinem (sodium) is a beta-lactamase-stable tricyclic β-lactam antibiotic with broad-spectrum activity against gram-positive and gram-negative bacteria. It is provided for research use and is supplied in solid forms and as a 10 mM solution in DMSO for in-vitro testing and antimicrobial susceptibility studies.
  • Beta-lactamase-stable tricyclic β-lactam antibiotic.
  • Broad-spectrum activity against gram-positive and gram-negative bacteria.
  • Available as solid material and as a 10 mM solution in DMSO.
  • Suitable for in-vitro antimicrobial susceptibility and resistance studies.
  • Provided with CAS and molecular weight reference data for documentation.
Catalog No. 50-002-39908